Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Adenosine A2aR/A2bR Alexa Fluor 405-conjugated Antibody, R&D Systems™
Mouse Monoclonal Antibody
Supplier: R&D Systems FAB94972V100UG
This item is not returnable.
View return policy
Description
Adenosine A2aR/A2bR Monoclonal specifically detects Adenosine A2aR/A2bR in Human samples. It is validated for Flow Cytometry.Specifications
Adenosine A2aR/A2bR | |
Monoclonal | |
Alexa Fluor 405 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Human | |
Purified |
Flow Cytometry | |
599743 | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
P29274 | |
NS0 mouse myeloma cell line transfected with human Adenosine A2aR, Met1-Ser412, Accession # P29274 | |
100 μg | |
Primary | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied. | |
IgG2b |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction