Learn More
Adenosine A2aR Mouse anti-Human, Alexa Fluor™ 405, Clone: 599717R, R&D Systems™
Mouse Monoclonal Antibody
Manufacturer:  R&D Systems FAB9497RV100UG
Catalog No. FAB9497RV10
Add to cart
This item is not returnable. View return policy
Description
Description
Adenosine A2aR Monoclonal antibody specifically detects Adenosine A2aR in Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
Adenosine A2aR
599717R
Flow Cytometry 0.25-1 μg/106 cells
P29274
A2aR, adenosine A2 receptor, Adenosine A2a R, adenosine A2a receptor, adenosine receptor A2a, adenosine receptor subtype A2a, ADORA 2, ADORA2, ADORA2A, hA2aR, RDC8
NS0 mouse myeloma cell line transfected with human Adenosine A2a R, Met1-Ser412, Accession # P29274
Protein A or G purified from cell culture supernatant
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied.
Monoclonal
Detects human Adenosine A2a R in direct ELISAs.
Flow Cytometry
Alexa Fluor 405
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide.
Purified, Supernatant, Conjugated
Mouse
IgG2a
100 μg
Primary
135
Human