Learn More
Abnova™ Adenosine Assay Kit (Fluorometric)
Adenosine Assay Kit (Fluorometric) is used to determine adenosine.
Manufacturer:  Abnova™ KA4547
Catalog No. 89-932-663
Add to cart
Description
Description
  • Suitable Sample: Plasma, urine
  • Calibration Range: 2 to 80 pmol
  • Reactivity: Mammals
  • Quality Control Testing: Standard Curve
Specifications
Specifications
Functional Study
1 Kit
Store the kit at -20°C, protected from light.
Kit