Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Adenoviral capsid protein VI Mouse anti-Virus, Clone: 9F10, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma MABF2196100UG
Description
Adenoviral capsid protein VI Monoclonal antibody specifically detects Adenoviral capsid protein VI in Virus samples. It is validated for Western Blotting, Immunofluorescence.Specifications
Adenoviral capsid protein VI | |
Monoclonal | |
1 mg/mL | |
Purified mouse monoclonal antibody IgG1 in buffer containing 0.1M Tris-Glycine, 0.15M NaCl, 0.05% sodium azide, pH7.4 with 0.05% sodium azide. | |
Recombinant purified adenovirus capsid protein VI. | |
100 μg | |
Primary | |
Stable for 1 year at 2-8°C from date of receipt. | |
IgG1 κ |
Western Blot, Immunofluorescence | |
9F10 | |
Unconjugated | |
Mouse | |
Protein G purified | |
RUO | |
Virus | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction