Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Adenoviral capsid protein VI Mouse anti-Virus, Clone: 9F10, MilliporeSigma™

Mouse Monoclonal Antibody

$153.40 - $387.00

Specifications

Antigen Adenoviral capsid protein VI
Clone 9F10
Concentration 1 mg/mL
Applications Western Blot, Immunofluorescence
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MABF2196100
View Documents
MilliporeSigma
MABF2196100UG
100 μg
Each for $387.00
Add to cart
 
MABF219625
View Documents
MilliporeSigma
MABF219625UG
25 μg
Each for $153.40
Add to cart
 
Description

Description

Adenoviral capsid protein VI Monoclonal antibody specifically detects Adenoviral capsid protein VI in Virus samples. It is validated for Western Blotting, Immunofluorescence.
Specifications

Specifications

Adenoviral capsid protein VI
1 mg/mL
Unconjugated
Mouse
Virus
Recombinant purified adenovirus capsid protein VI.
Monoclonal
Protein G purified
9F10
Western Blot, Immunofluorescence
Purified
RUO
Purified mouse monoclonal antibody IgG1 in buffer containing 0.1M Tris-Glycine, 0.15M NaCl, 0.05% sodium azide, pH7.4 with 0.05% sodium azide.
IgG1κ
Primary
Stable for 1 year at 2-8°C from date of receipt.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit