Learn More
Adenovirus Hexon Mouse anti-Adenovirus, Virus, FITC, Clone: 7F1, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP211639F
Catalog No. NBP211639F
Add to cart
Description
Description
Adenovirus Hexon Monoclonal antibody specifically detects Adenovirus Hexon in Adenovirus, Virus samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin).
Specifications
Specifications
Adenovirus Hexon
7F1
ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin
Purified
Mouse
IgG2a
0.1 ml
Store at 4C in the dark.
Monoclonal
ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
FITC
PBS with 0.05% Sodium Azide
Hexon protein; Late protein 2; PII
Hexon antigen of human adenovirus.
Protein A purified
RUO
Primary
Adenovirus, Virus