Learn More
Adenovirus Mouse anti-Adenovirus, Clone: CF302, Abnova™
Mouse Monoclonal Antibody
Manufacturer:  Abnova Corporation MAB13488
Catalog No. 89-033-453
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Mouse monoclonal antibody raised against adenovirus.
  • High specificity
  • High reactivity
Specifications
Specifications
Adenovirus
CF302
Mouse monoclonal antibody raised against adenovirus.
In PBS, pH 7.4 (0.09% sodium azide).
Mouse
IgG2a
1 mg
Store at 4°C.
Monoclonal
Adenovirus
ELISA, Immunohistochemistry, Lateral Flow
Unconjugated
ELISA Immunohistochemistry Lateral-Flow Immunochromatographic Assay The optimal working dilution should be determined by the end user.
Liquid
Native purified adenovirus hexon protein.
Protein A purification
RUO
Primary
This antibody reacts with hexon protein.
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Research Use Only (RUO)