Learn More
Adenovirus Mouse anti-Adenovirus, Clone: HM057, Abnova™
Mouse Monoclonal Antibody
Manufacturer:  Abnova Corporation MAB13492
Catalog No. 89-033-457
Add to cart
Description
Description
Mouse monoclonal antibody raised against adenovirus.
  • High specificity
  • High reactivity
Specifications
Specifications
Adenovirus
HM057
Mouse monoclonal antibody raised against adenovirus.
In PBS, pH 7.0 (0.02% sodium azide).
Mouse
IgG1
RUO
Primary
Adenovirus
Lateral Flow
Unconjugated
Lateral-Flow Immunochromatographic Assay The optimal working dilution should be determined by the end user.
Liquid
Purified human adenovirus.
1 mg
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Research Use Only (RUO)