Learn More
Adenovirus Mouse anti-Adenovirus, Clone: J312, Abnova™
Mouse Monoclonal Antibody
Manufacturer:  Abnova Corporation MAB13493
Catalog No. 89-033-458
Add to cart
Description
Description
Mouse monoclonal antibody raised against adenovirus.
  • High specificity
  • High reactivity
Specifications
Specifications
Adenovirus
J312
Mouse monoclonal antibody raised against adenovirus.
In 10 mM PBS, pH 7.2 (0.09% sodium azide).
Mouse
IgG1
1 mg
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
Adenovirus
ELISA, Immunofluorescence, Lateral Flow
Unconjugated
ELISA Immunofluorescence Lateral-Flow Immunochromatographic Assay The optimal working dilution should be determined by the end user.
Liquid
Native purified adenovirus hexon protein.
Protein A purification
RUO
Primary
This antibody reacts with hexon protein. Reacts with adenovirus serotypes 1,2,3,4,5,6,7,8, 9,10,11,12, 14,16,18,20,26,31,34,35,36,37,40 and 41.
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Research Use Only (RUO)