Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Adenovirus Mouse anti-Virus, Clone: 8C4, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 6 publications
$152.22 - $344.00
Specifications
Antigen | Adenovirus |
---|---|
Clone | 8C4 |
Dilution | Flow Cytometry 1:10-1:1000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500, Immunodiffusion, Sandwich ELISA 1:100-1:2000, CyTOF-ready, Block/Neutralize |
Applications | Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunodiffusion, ELISA, CyTOF, Neutralization |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB600413SS
|
Novus Biologicals
NB600413SS |
0.05 mg |
Each for $152.22
|
|
NB600413
|
Novus Biologicals
NB600413 |
0.2 mg |
Each for $344.00
|
|
Description
Adenovirus Monoclonal specifically detects Adenovirus in Adenovirus, Virus samples. It is validated for ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunodiffusion, Sandwich ELISA, Lateral Flow Assay, Block/Neutralize.Specifications
Adenovirus | |
Flow Cytometry 1:10-1:1000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500, Immunodiffusion, Sandwich ELISA 1:100-1:2000, CyTOF-ready, Block/Neutralize | |
Monoclonal | |
Purified | |
RUO | |
Adenovirus, Virus | |
Hexon antigen of human adenovirus. | |
Primary | |
Store at 4C. Do not freeze. |
8C4 | |
Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunodiffusion, ELISA, CyTOF, Neutralization | |
Unconjugated | |
Mouse | |
Cell Biology, Stem Cell Markers, Virology Bacteria and Parasites | |
Adenovirus | |
IgG2a | |
Protein A purified | |
Adenovirus hexon specific. The is directed against a genus-specific hexon antigen, which is present in all human serotypes. This detects a different epitope than the monoclonal, NB200-425. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title