Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Epredia™ Lab Vision™ Adenovirus Type 2 and 5 Ab-5 Mouse Monoclonal Antibody, 200μg/mL, BSA and azide
Obtain accurate, reproducible results in immunohistochemistry, western blotting, immunofluorescence, and immunoprecipitation experiments.
Supplier: Epredia™ MS1069PABX
Description
Adenovirus Ab-5 detects adenovirus serogroups 2 and 5. The adenovirus early gene product E1A is a potent stimulator of cellular proliferation, which when overexpressed can overcome the growth-inhibitory effects of TGF-β. The E1A region encodes a series of related proteins (35-46kDa) with multifuctional capabilities and form a specific complex with the retinoblastoma tumor suppressor gene product (Rb protein). The E1A and E1B regions together comprise the transforming region of adenovirus. While expression of E1A alone is sufficient to immortalize primary cells, complete transformation requires the additional expression of the E1B region. Several conserved regions of E1A are similar to portions of other viral oncoproteins like the HPV-16 and HPV-18 E7 and SV40 large T antigen.Host Species: Mouse
Clone: M58 + M73
Isotype: IgG2a/κ + IgG2a/κ
Species Reactivity: Adenovirus.
Immunogen: Adenovirus
Positive Control: 293 cells or adenovirus infected tissues
Cellular Localization: Nuclear.
Ab-5 is excellent for staining of formalin-fixed, paraffin-embedded tissues.
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting
- Immunofluorescence
- Immunoprecipitation (Native verified)
Note:
USA: RUO; Int'l: RUO
Specifications
Adenovirus Type 2 and 5 Ab-5 | |
Monoclonal | |
1mg/mL | |
Mouse | |
200 μL | |
Infectious Disease | |
Adenovirus | |
IgG2a κ |
Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
M58 + M73 | |
Unconjugated | |
Adenovirus | |
RUO | |
Primary | |
Concentrated |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction