Learn More
Invitrogen™ Adenovirus Type 7 Monoclonal Antibody (AF-1)
Description
MA1-7600 detects Adenovirus from virus samples. MA1-7600 has been successfully used in ELISA, Western blot and Immunofluorescence procedures. The MA1-7600 immunogen is a purified Ad7 virus.
Specifications
Specifications
| Antigen | Adenovirus Type 7 |
| Applications | ELISA, Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | AF-1 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.01% sodium azide; pH 7.2 |
| Gene Alias | Adeno Virus; Adeno Virus 7; AdV; AdV-7; HADV; HADV-7 |
| Host Species | Mouse |
| Immunogen | Purified Ad7 virus. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.