Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Adenovirus 2 Early E1A Mouse anti-Adenovirus, Virus, Clone: M73, Novus Biologicals
Mouse Monoclonal Antibody
Manufacturer: Novus Biologicals NBP266587
Description
Adenovirus 2 Early E1A Monoclonal antibody specifically detects Adenovirus 2 Early E1A in Adenovirus, Virus samples. It is validated for Immunocytochemistry, Immunofluorescence, Immunoprecipitation.Specifications
Adenovirus2EarlyE1A | |
M73 | |
Immunocytochemistry/Immunofluorescence 1:10 - 1:500, Immunoprecipitation 1:10-1:500 | |
Purified | |
Full length recombinant adenovirus 2 E1A protein | |
Ammonium sulfate precipitation | |
Store at -20°C. Avoid freeze-thaw cycles. | |
Monoclonal |
Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
Unconjugated | |
PBS with 0.08% Sodium Azide | |
Mouse | |
IgG2a | |
100μg | |
Primary | |
Adenovirus, Virus |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title