Learn More
BioVision IncSupplier Diversity Partner Adenylate Kinase (AK) Activity Assay Kit (Colorimetric/Fluorometric): Adenylate Kinase (AK) Activity Assay Kit (Colorimetric/Fluorometric): Rapid & Convenient Assay for Detecting AK Activity in Biological samples and Purified enzyme. 100 Assays. DFS Item
Manufacturer:  BioVision IncSupplier Diversity Partner K350100
Adenylate Kinase (AK) Activity Assay Kit (Colorimetric/Fluorometric): Adenylate Kinase (AK) (EC 2.7.4.3) is anenzyme involved in energy metabolism and homeostasis of cellular adenine nucleotide ratios in different intracellular compartments. The enzyme is found in the nucleus, cytosol, or mitochondria (intermembrane space or matrix) of various kinds of tissues. Adenylate kinase acts on two molecules of ADP to generate ATP and AMP. Nine isoforms of adenylate kinase have been identified. Erythrocyte adenylate kinase deficiency is associated with hemolytic anemia. BioVisions AK Activity Assay kit can kinetically measure Adenylate Kinase activity by detecting adenosine triphosphate (ATP) generated from adenosine diphosphate (ADP) as a substrate. ATP is detected via a multi-step reaction, resulting in the generation of an intermediate that reacts with the Adenylate Kinase Probe forming an end product that can be measured colorimetrically (OD 570 nm) or fluorometrically (Ex/Em = 535/587 nm
Catalog No. 50-111-9713