Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Adenylate Cyclase 3 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Supplier:  Thermo Scientific PA131191

Catalog No. PIPA131191

Item Discontinued This item has been discontinued by the supplier and is no longer available. Please reach out to your sales representative.


Description

Description

PA1-31191 detects ADCY3 from rat, canine, human, mouse samples. PA1-31191 has been successfully used in Western blot applications. The PA1-31191 immunogen is: The antibodies were generated against a synthetic peptide sequence (unfortunately, the amino acid sequence is considered to be commercially sensitive) unique to PAC3 protein. All peptides were amidated before conjugation. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

This gene encodes adenylyl cyclase 3 which is a membrane-associated enzyme and catalyzes the formation of the secondary messenger cyclic adenosine monophosphate (cAMP). This protein appears to be widely expressed in various human tissues and may be involved in a number of physiological and pathophysiological metabolic processes.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Adenylate Cyclase 3
Polyclonal
Unconjugated
O60266, Q8VHH7, P21932
ADCY3
The antibodies were generated against a synthetic peptide sequence (unfortunately, the amino acid sequence is considered to be commercially sensitive) unique to PAC3 protein. All peptides were amidated before conjugation.
100 μg
Primary
Canine, Human, Mouse, Rat
Antibody
IgG
Western Blot
0.75 mg/mL
ADCY3
AC3, KIAA0511
Rabbit
Antigen affinity chromatography
RUO
104111, 109, 482994, 64508
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Recommendations

Recommendations

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit