Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Adenylate Cyclase 9 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5101976
Description
Antibody detects endogenous levels of total ADCY9.
Adenylate cyclase is a membrane bound enzyme that catalyses the formation of cyclic AMP from ATP. It is regulated by a family of G protein-coupled receptors, protein kinases, and calcium. The type 9 adenylyl cyclase is a widely distributed adenylyl cyclase, and it is stimulated by beta-adrenergic receptor activation but is insensitive to forskolin, calcium, and somatostatin.
Specifications
Adenylate Cyclase 9 | |
Polyclonal | |
Unconjugated | |
ADCY9 | |
AC9; ACIX; ACtp10; Adcy9; adenylate cyclase 9; adenylate cyclase type 9; Adenylate cyclase type IX; adenylyl cyclase 9; Adenylyl cyclase type 10; ATP pyrophosphate-lyase 9; AW125421; D16Wsu65e; KIAA0520; mKIAA0520; type IX ATP pyrophosphate-lyase | |
Rabbit | |
Affinity chromatography | |
RUO | |
115 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
O60503 | |
ADCY9 | |
A synthesized peptide derived from human ADCY9(Accession O60503), corresponding to amino acid residues S944-W994. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction