Learn More
Adenylate Kinase 1 Antibody (OTI4A1), DyLight 755, Novus Biologicals™
SDP
Mouse Monoclonal Antibody
Supplier:  Novus Biologicals NBP270123IR
Catalog No. NBP270123IR
Add to Cart
Description
Description
Adenylate Kinase 1 Monoclonal specifically detects Adenylate Kinase 1 in Human, Mouse, Rat, Monkey samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
Adenylate Kinase 1
Monoclonal
DyLight 755
50mM Sodium Borate with 0.05% Sodium Azide
AK1
Full length recombinant protein of human Adenylate Kinase 1 (NP_000467) produced in HEK293T cells.
0.1 mL
Primary
Human, Mouse, Rat, Monkey
IgG1
Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence
OTI4A1
Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence
adenylate kinase 1, adenylate kinase isoenzyme 1, AK 1, ATP-AMP transphosphorylase 1, EC 2.7.4, EC 2.7.4.3, Myokinase
Mouse
Affinity Purified
metabolism, Signal Transduction
203.0
Store at 4°C in the dark.
Product Suggestions
Product Suggestions