Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Adenylate Kinase 1 Mouse anti-Human, Monkey, PE-Cyanine7, Clone: OTI4A1, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP270123PECY7

Catalog No. N270123PEC7


Add to cart

Description

Description

Adenylate Kinase 1 Monoclonal antibody specifically detects Adenylate Kinase 1 in Human, Monkey samples. It is validated for Flow Cytometry.
Specifications

Specifications

Adenylate Kinase 1
OTI4A1
Flow Cytometry
Affinity Purified, Conjugated
AK1
Full length recombinant protein of human Adenylate Kinase 1 (NP_000467) produced in HEK293T cells.
Immunogen affinity purified
Store at 4°C in the dark. Do not freeze.
Monoclonal
Human, Monkey
Flow Cytometry
PE-Cyanine7
PBS with 0.05% Sodium Azide
adenylate kinase 1, adenylate kinase isoenzyme 1, AK 1, ATP-AMP transphosphorylase 1, EC 2.7.4, EC 2.7.4.3, Myokinase
Mouse
IgG1
0.1 mL
Primary
203.0
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit