Learn More
Adenylate Kinase 1 Mouse anti-Human, Non-human primate, Clone: OTI19D1, Invitrogen™
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MA524905
Catalog No. PIMA524905
Add to cart
Description
Description
Adenylate kinase is an enzyme involved in regulating the adenine nucleotide composition within a cell by catalyzing the reversible transfer of phosphate group among adinine nucleotides. Three isozymes of adenylate kinase have been identified in vertebrates, adenylate isozyme 1 (AK1), 2 (AK2) and 3 (AK3). AK1 is found in the cytosol of skeletal muscle, brain and erythrocytes, whereas AK2 and AK3 are found in the mitochondria of other tissues including liver and heart. AK1 was identified because of its association with a rare genetic disorder causing nonspherocytic hemolytic anemia where a mutation in the AK1 gene was found to reduce the catalytic activity of the enzyme.
Specifications
Specifications
Adenylate Kinase 1
Monoclonal
1.2 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
P00568
AK 1, ATP-AMP transphosphorylase 1, adenylate kinase isoenzyme 1, myokinase
Mouse
IgG2b
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
203, 706338
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot
OTI19D1
Unconjugated
AK1
Liquid
AK1
Full length human recombit protein of AK1 produced in HEK293T cell
Affinity Chromatography
RUO
Antibody
Monoclonal
Human, Non-human primate