Learn More
Adenylate Kinase 4 Mouse anti-Canine, Human, Non-human primate, Clone: OTI2B12, Invitrogen™
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MA525640
Catalog No. PIMA525640
Add to cart
Description
Description
This gene encodes a member of the adenylate kinase family of enzymes. The encoded protein is localized to the mitochondrial matrix. Adenylate kinases regulate the adenine and guanine nucleotide compositions within a cell by catalyzing the reversible transfer of phosphate group among these nucleotides. Five isozymes of adenylate kinase have been identified in vertebrates. Expression of these isozymes is tissue-specific and developmentally regulated. A pseudogene for this gene has been located on chromosome 17. Three transcript variants encoding the same protein have been identified for this gene. Sequence alignment suggests that the gene defined by NM_013410, NM_203464, and NM_001005353 is located on chromosome 1.
Specifications
Specifications
Adenylate Kinase 4
Monoclonal
0.42 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
P27144
RP4-686B20.1, AK3, AK3L1, AK3L2, ATP-AMP transphosphorylase, GTP:AMP phosphotransferase, adenylate kinase 3-like 1, adenylate kinase isoenzyme 4, mitochondrial, mitochondrial adenylate kinase-3, nucleoside-triphosphate-adenylate kinase
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
205, 489554, 698560
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
OTI2B12
Unconjugated
AK4
Liquid
AK4
Full length human recombit protein of AK4 produced in HEK293 cell
Affinity Chromatography
RUO
Antibody
Monoclonal
Canine, Human, Non-human primate