Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Adenylosuccinate Lyase Antibody (OTI2D10), PE, Novus Biologicals™
SDP

Mouse Monoclonal Antibody

Supplier:  Novus Biologicals NBP270101PE

Catalog No. NBP270101PE


Add to Cart

Description

Description

Adenylosuccinate Lyase Monoclonal specifically detects Adenylosuccinate Lyase in Human, Mouse, Rat, Canine, Primate, Monkey samples. It is validated for Flow Cytometry.
Specifications

Specifications

Adenylosuccinate Lyase
Monoclonal
PE
PBS with 0.05% Sodium Azide
ADSL
Full length recombinant protein of human Adenylosuccinate Lyase (NP_000017) produced in HEK293T cells.
0.1 mL
Primary
Human, Mouse, Rat, Canine, Primate, Monkey
IgG1
Flow Cytometry
OTI2D10
Flow Cytometry
adenylosuccinase, adenylosuccinate lyase, AMPS, ASase, ASL, EC 4.3.2, EC 4.3.2.2
Mouse
Affinity Purified
Core ESC Like Genes, Stem Cell Markers
158.0
Store at 4°C in the dark.
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit