Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ADFP Mouse anti-Human, Clone: ADFP/1494, Abnova™

Mouse Monoclonal Antibody

Manufacturer:  Abnova Corporation MAB14487

Catalog No. 89-034-570


Add to cart

Description

Description

Mouse monoclonal antibody raised against partial recombinant human ADFP.

  • High specificity
  • High reactivity
Specifications

Specifications

ADFP
ADFP/1494
Mouse monoclonal antibody raised against partial recombinant human ADFP.
In 10 mM PBS (0.05% BSA, 0.05% sodium azide).
Liquid
ADFP
Recombinant protein corresponding to amino acids 249-376 of human ADFP.
Protein A/G purification
RUO
Primary
123
Immunofluorescence, Immunohistochemistry (PFA fixed)
Unconjugated
Immunofluorescence (0.5-1 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (2-4 ug/mL) The optimal working dilution should be determined by the end user.
Q99541
ADRP|MGC10598
Mouse
IgG2b, kappa
100 ug
Store at 4°C.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only (RUO)