Learn More
BioVision IncSupplier Diversity Partner Alcohol Dehydrogenase Activity Colorimetric Assay Kit: Alcohol Dehydrogenase (ADH) Activity Assay Kit: Colorimetric Assay for Detecting ADH Activity in Cells, Tissues, Serum etc. Simple, Sensitive & Accurate. Detection Limit: 0.01 mU. 100 Assays. DFS Item
Manufacturer:  BioVision IncSupplier Diversity Partner K787100
Alcohol dehydrogenase (Alcohol DH, ADH) (EC 1.1.1.1) is a group of seven dehydrogenase enzymes that occur in many organisms and facilitate the interconversion between alcohols and aldehydes or ketones with the reduction of NADᶧ to NADH. In humans and many other animals, they serve to break down alcohols which could otherwise be toxic; in yeast and many bacteria, some alcohol dehydrogenases catalyze the opposite reaction as part of fermentation. BioVision’s Alcohol Dehydrogenase Assay Kit provides a convenient tool for sensitive detection of the Alcohol DH in a variety of samples. In the assay Alcohol DH will utilize isopropanol as a substrate leading to a proportional color development. The activity of ADH can be easily quantified colorimetrically (λ = 450 nm). This assay detects ADH activity as low as 0.01 mU in samples. BioVision charges an Ice pack and Insulated Container fee of $25.00 for each order.
Catalog No. 50-111-9905