Learn More
ADH1B Rabbit anti-Human, Polyclonal, Abnova™
Rabbit Polyclonal Antibody
Manufacturer:  Abnova Corporation PAB29951
Catalog No. 89-033-239
Add to cart
Description
Description
Rabbit polyclonal antibody raised against synthetic peptide of human ADH1B.
  • High specificity
  • High reactivity
Specifications
Specifications
ADH1B
Unconjugated
Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (4-8 ug/mL) Western Blot (1.25 ug/mL) The optimal working dilution should be determined by the end user.
NM_000668
ADH2|DKFZp686C06125
Rabbit
Protein A purification
RUO
Primary
125
Immunohistochemistry (PFA fixed), Western Blot
Rabbit polyclonal antibody raised against synthetic peptide of human ADH1B.
In PBS (2% sucrose, 0.09% sodium azide).
Liquid
ADH1B
A synthetic peptide corresponding to C-terminus of human ADH1B.
100 uL
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Polyclonal
Human
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Research Use Only (RUO)