Learn More
Aviva Systems Biology Corp ADH5 Antibody - middle region (ARP51767_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP51767_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ADH5. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ADH5. Alternate names: ADH-3, ADHX, FDH, FALDH, GSNOR, GSH-FDHApplicationWB. Species reactivity: Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-62845