Learn More
Abnova™ ADH6 293T Cell Transient Overexpression Lysate (Denatured)
Human overexpression lysate
Manufacturer:  Abnova™ H00000130T01
Catalog No. 89-932-688
Add to cart
Description
Description
Specifications
Specifications
293T
BC039065.1
pCMV-ADH6 full-length
100 μL
Human
ADH6 293T Cell Transient Overexpression Lysate(Denatured)
Theoretical M.W. 32.56kDa
Transient overexpression cell lysate was tested with anti-ADH6 antibody (Mfr. No. H00000130-B01) by Western Blot.
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
Western Blot