Brady Corporation ABSORBENT MAT AD15100 1/RL

Manufacturer: Brady Corporation AD15100

Catalog No. 19-103-108

  Check Availability
Add to Cart

    Specifications

    Length (English) 100 ft.
    For Use With (Application) Walkways, areas of heavy traffic and in place of traditional rugs
    Material Polypropylene
    Item Description Needle-punched polypropylene with adhesive backing
    Width (English) 15 in.
    Capacity 11 gal.
    Dimensions (L x W) 100 ft. x 15 in.
    Packaging Quantity 1
    Quantity 1/Roll
    Color Black
    Absorbancy 8 gl/gs

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit