Learn More
Thermo Scientific™ Adhesive Plate Seals Available on GSA/VA Contract for Federal Government customers only.
Thermo Scientific™ Gas Permeable Adhesive Seal provides a simple and effective way of venting cultures while providing a barrier against contaminating bacteria.
$155.70 - $156.50
Products 3
Catalog Number Mfr. No. For Use With (Application) Price Quantity & Availability  
Catalog Number Mfr. No. For Use With (Application) Price Quantity & Availability  
AB-0580 Available on GSA/VA Contract for Federal Government customers only.
View Documents
Thermo Scientific™
AB0580
For crystallography and genotyping Case of 100 for $156.50
Add to cart
AB-0718 Available on GSA/VA Contract for Federal Government customers only.
View Documents
Thermo Scientific™
AB0718
Ventilating and storing bacterial/cell cultures Case of 50 for $155.70
Add to cart
AB-1450
View Documents
Thermo Scientific™
AB1450
Long- or short-term sample storage This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description
Description
Adhesive Plate Seals are more convenient and cost effective than other sealing methods, and are available in a wide range of styles to suit any application. Thermo Scientific adhesive seals do not leave behind a sticky residue when the seal is removed, allowing the plate to be sealed multiple times.
Thermo Scientific Adhesive Plate Seals provide an effective seal. They feature an easy-removal non-adhesive tab and can withstand temperatures between -40°C to 80°C.
Features of Thermo Scientific Adhesive Plate Seals
  • Non-adhesive tab for easy removal of seal
  • Broad temperature range: -40°C to 80°C
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.