Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ADHFE1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA550541

Catalog No. PA550541


Add to cart

Description

Description

The antibody detects endogenous levels of total ADHFE1 protein.

The ADHFE1 gene encodes hydroxyacid-oxoacid transhydrogenase (EC 1.1.99.24), which is responsible for the oxidation of 4-hydroxybutyrate in mammalian tissues (Kardon et al., 2006 [PubMed 16616524]).
Specifications

Specifications

ADHFE1
Polyclonal
Unconjugated
ADHFE1
Liquid
ADHFE1
Synthetic peptide corresponding to a region derived from internal residues of human alcohol dehydrogenase, iron containing, 1
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Immunohistochemistry (Paraffin)
1.6 mg/mL
PBS with 50% glycerol and 0.05% sodium azide; pH 7.3
Q8IWW8, Q8R0N6, Q4QQW3
HOT, ADH8, HMFT2263
Rabbit
IgG
100 μL
-20°C
Primary
137872, 362474, 76187
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit