Learn More
ADHFE1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 193591AP
Catalog No. 50-173-2388
Add to cart
Description
Description
The ADHFE1 gene encodes hydroxyacid-oxoacid transhydrogenase (EC 1.1.99.24), which is responsible for the oxidation of 4-hydroxybutyrate in mammalian tissues (Kardon et al., 2006 ).
Specifications
Specifications
ADHFE1
0.26 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q4QQW3, Q8IWW8, Q8R0N6
ADH8, ADHFE1, HMFT2263, HOT
Rabbit
IgG
150 μL
-20°C
Primary
137872, 362474, 76187
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Unconjugated
ADHFE1
Liquid
Adhfe1
ADHFE1 Fusion Protein Ag6803
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat