Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ADI1 Mouse anti-Human, Clone: OTI3H6, liquid, TrueMAB™ Non-distribution product as customer accommodation.

Mouse Monoclonal Antibody

Manufacturer:  OriGeneSupplier Diversity Partner TA501755

Catalog No. 50-167-2873


Add to cart

Description

Description

This gene encodes an enzyme that belongs to the aci-reductone dioxygenase family of metal-binding enzymes, which are involved in methionine salvage. This enzyme may regulate mRNA processing in the nucleus, and may carry out different functions depending on its localization. A pseudogene that is located on chromosome 20 has been defined for this gene.
Specifications

Specifications

ADI1
Monoclonal
0.47 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q9BV57
APL1, ARD, Fe-ARD, HMFT1638, MTCBP1, Ni-ARD, SIPL, mtnD
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
55256
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot
OTI3H6
Unconjugated
ADI1
Liquid
ADI1
Full length human recombit protein of human ADI1 produced in HEK293T cell.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit