Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ADIG Polyclonal Antibody
Goat Polyclonal Antibody
Supplier: Invitrogen™ PA5143174
This item is not returnable.
View return policy
Description
This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:2,000. Preliminary analysis showed no signal in Western Blot.

Specifications
ADIG | |
Polyclonal | |
Unconjugated | |
ADIG | |
ADIG; adipogenesis associated; adipogenin; SMAF1; small adipocyte factor 1 (SMAF1) | |
Goat | |
Ammonium sulfate precipitation | |
RUO | |
149685 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
Q0VDE8 | |
ADIG | |
Peptide with sequence SQESKEDDSDLC (aa 41-52). | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction