Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Adipic Acid, FCC, 99.6-101%, Spectrum™

Click to view available options
Quantity:
25 g
100 g
500 g
3 kg
12 kg
45 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
124-04-9
C6H10O4
146.14
WNLRTRBMVRJNCN-UHFFFAOYSA-N
hexanedioic acid
OC(=O)CCCCC(O)=O
124-04-9
C6H10O4
146.14
WNLRTRBMVRJNCN-UHFFFAOYSA-N
hexanedioic acid
OC(=O)CCCCC(O)=O

AD130, 124-04-9, C6H10O4

Spectrum™ Adipic Acid, FCC is used as a food ingredient as a flavorant and gelling aid and as an acidulant in baking powders. Spectrum Chemical offers over 300 Food Grade (FCC) chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
CAS Max % 1
Percent Purity 99.6 to 101%
Ignition Residue 2e-005
Lead (Pb) 2 mg/kg
Packaging Poly Pail
Water 0.002
Quantity 3 kg