Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Adipic dihydrazide, 98%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
500g
Packaging:
Plastic bottle
1071-93-8
C6H14N4O2
174.2
MFCD00007614
IBVAQQYNSHJXBV-UHFFFAOYSA-N
adipic dihydrazide,adipohydrazide,adipic acid dihydrazide,hexanedioic acid, dihydrazide,adipyl hydrazide,hexanediohydrazide,adipic acid, dihydrazide,adipodihydrazide,hexanedioic acid, 1,6-dihydrazide,unii-vk98i9yw5m
66117
hexanedihydrazide
C(CCC(=O)NN)CC(=O)NN
1071-93-8
C6H14N4O2
174.2
MFCD00007614
IBVAQQYNSHJXBV-UHFFFAOYSA-N
adipic dihydrazide,adipohydrazide,adipic acid dihydrazide,hexanedioic acid, dihydrazide,adipyl hydrazide,hexanediohydrazide,adipic acid, dihydrazide,adipodihydrazide,hexanedioic acid, 1,6-dihydrazide,unii-vk98i9yw5m
66117
hexanedihydrazide
C(CCC(=O)NN)CC(=O)NN
Color White to Yellow
Melting Point 178.0°C to 182.0°C
Infrared Spectrum Authentic
Packaging Plastic bottle
Quantity 25g
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula H2NNHCO(CH2)4CONHNH2
Beilstein 02, I, 277
Solubility Information Solubility in water: soluble. Other solubilities: soluble in acetic acid,slightly soluble in acetone,insoluble in ethanol,ether and benzene
Formula Weight 174.2
Physical Form Crystalline Powder
Percent Purity 98%
Chemical Name or Material Adipic dihydrazide
Show More Show Less

GHS H Statement
May cause an allergic skin reaction.
Toxic to aquatic life with long lasting effects.

GHS P Statement
Wear protective gloves/protective clothing.
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation or rash occurs: Get medical advice/attention.

GHS Signal Word: Warning

EINECSNumber : 213-999-5