Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Adiponectin/Acrp30 Mouse anti-Human, PE-Cyanine7, Clone: OTI1H1, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP271422PECY7

Catalog No. N271422PEC7


Add to cart

Description

Description

Adiponectin/Acrp30 Monoclonal antibody specifically detects Adiponectin/Acrp30 in Human samples. It is validated for Flow Cytometry.
Specifications

Specifications

Adiponectin/Acrp30
OTI1H1
Flow Cytometry
Affinity Purified, Conjugated
ADIPOQ
Recombinant protein fragment corresponding to amino acids 19-244 of human Adiponectin/Acrp30 (NP_004788) produced in HEK293T cells.
Immunogen affinity purified
Diabetes Research, Lipid and Metabolism
Primary
9370.0
Flow Cytometry
PE-Cyanine7
PBS with 0.05% Sodium Azide
ACDC, ACRP30ADPN, adipocyte, C1Q and collagen domain containing, adiponectin, adiponectin, C1Q and collagen domain containing, AdipoQ, ADIPQTL1, apM-1, APM1APM-1, C1q and collagen domain-containing protein, GBP28apM1, Gelatin-binding protein
Mouse
IgG1
0.1 mL
Store at 4°C in the dark. Do not freeze.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit