Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Adiponectin/Acrp30, Mouse anti-Mouse, Clone: MADI 1147, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP280030100UG
Description
Adiponectin/Acrp30 Monoclonal specifically detects Adiponectin/Acrp30 in Mouse samples. It is validated for Western Blot, ELISA, Immunoprecipitation.Specifications
Adiponectin/Acrp30 | |
Monoclonal | |
1 mg/mL | |
Western Blot, ELISA, Immunoprecipitation | |
ACDC, ACRP30ADPN, adipocyte, C1Q and collagen domain containing, adiponectin, adiponectin, C1Q and collagen domain containing, AdipoQ, ADIPQTL1, apM-1, APM1APM-1, C1q and collagen domain-containing protein, GBP28apM1, Gelatin-binding protein | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Recognizes mouse Adiponectin/Acrp30. Detects a band of ∽30kDa by Western blot. Weakly cross-reacts with human and rat Adiponectin/Acrp30. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG1 κ |
Western Blot, ELISA, Immunoprecipitation | |
MADI 1147 | |
Unconjugated | |
0.2 um-filtered solution in PBS, pH 7.4 | |
ADIPOQ | |
Recombinant mouse Adiponectin/Acrp30. | |
100 μg | |
Diabetes Research, Lipid and Metabolism | |
9370 | |
Mouse | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title