Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Adiponectin/Acrp30, Mouse anti-Mouse, Clone: MADI 1147, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP28003050UG

 View more versions of this product

Catalog No. NB013138


Add to cart

Description

Description

Adiponectin/Acrp30 Monoclonal specifically detects Adiponectin/Acrp30 in Mouse samples. It is validated for Western Blot, ELISA, Immunoprecipitation.
Specifications

Specifications

Adiponectin/Acrp30
MADI 1147
Unconjugated
0.2 um-filtered solution in PBS, pH 7.4
ACDC, ACRP30ADPN, adipocyte, C1Q and collagen domain containing, adiponectin, adiponectin, C1Q and collagen domain containing, AdipoQ, ADIPQTL1, apM-1, APM1APM-1, C1q and collagen domain-containing protein, GBP28apM1, Gelatin-binding protein
Mouse
IgG1 Kappa
50 ug
Diabetes Research, Lipid and Metabolism
Primary
9370
Mouse
Western Blot, ELISA, Immunoprecipitation
1 mg/mL
Western Blot, ELISA, Immunoprecipitation
Purified
ADIPOQ
Recombinant mouse Adiponectin/Acrp30.
Protein G purified
RUO
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Monoclonal
Recognizes mouse Adiponectin/Acrp30. Detects a band of ∽30kDa by Western blot. Weakly cross-reacts with human and rat Adiponectin/Acrp30.
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit