Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Adiponectin/Acrp30, Mouse anti-Rat, Clone: RADI 264, Novus Biologicals™

Mouse Monoclonal Antibody

$240.50 - $314.00

Specifications

Antigen Adiponectin/Acrp30
Clone RADI 264
Applications Western Blot, ELISA
Host Species Mouse
Target Species Rat
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB013151
View Documents
Novus Biologicals
NBP280037100UG
100 ug
Each for $314.00
Add to cart
 
NB013152
View Documents
Novus Biologicals
NBP28003750UG
50 ug
Each for $240.50
Add to cart
 
Description

Description

Adiponectin/Acrp30 Monoclonal antibody specifically detects Adiponectin/Acrp30 in Rat Samples. It is validated for Western Blot, ELISA.
Specifications

Specifications

Adiponectin/Acrp30
Western Blot, ELISA
Rat
Primary
RADI 264
Mouse
Monoclonal
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit