Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Adiponectin/Acrp30 Rabbit anti-Human, Mouse, Rat, Alexa Fluor™ 700, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NB10065810AF700

Catalog No. N1065810A70


Add to cart

Description

Description

Adiponectin/Acrp30 Polyclonal antibody specifically detects Adiponectin/Acrp30 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin).
Specifications

Specifications

Adiponectin/Acrp30
Alexa Fluor™ 700
50 mM sodium borate with 0.05% sodium azide
ACDC, ACRP30ADPN, adipocyte, C1Q and collagen domain containing, adiponectin, adiponectin, C1Q and collagen domain containing, AdipoQ, ADIPQTL1, apM-1, APM1APM-1, C1q and collagen domain-containing protein, GBP28apM1, Gelatin-binding protein
Rabbit
IgG
0.1 mL
Store at 4°C in the dark.
Polyclonal
This recognises an epitope within the C-terminal region (CT) of the 244 amino acid protein Adiponectin.
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Flow (Intracellular)
Affinity Purified, Conjugated
ADIPOQ
A synthetic peptide made to a C-terminal sequence of human Adiponectin (between amino acids 200-244) [UniProt Q15848]
Immunogen affinity purified
Diabetes Research, Lipid and Metabolism
Primary
9370.0
Human, Mouse, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit