Learn More
BioVision IncSupplier Diversity Partner Adiponectin (mouse) Elisa Assay Kit: Mouse Adiponectin ELISA Assay Kit: Sensitive & Convenient Assay for Quantitative Determination of Adiponectin in Mouse Serum, Plasma or Various Tissue or Cell Culture supernatants. 100 Assays. DFS Item
Manufacturer:  BioVision IncSupplier Diversity Partner K4902100
Adipose tissue secretes a number of biologically active soluble factors (collectively named adipocytokines) that regulate glucose and fatty acid metabolism. Measurement of serum adiponectin levels gives us important information on the role of adiponectin in regulation of glucose and/or lipid metabolism. This Mouse Adiponectin ELISA Kit is an enzyme-linked immunosorbent assay (ELISA) for quantitative determination of adiponectin in mouse serum, plasma or various tissue or cell culture supernatants. In the assay, monoclonal antibody specific for mouse adiponectin has been pre-coated onto 96 well microplate. Standards and samples are pipetted into the wells and adiponectin present is bound by immobilized antibody. The bound adiponectin is then captured by anti-mouse adiponectin polyclonal antibody. With HRP conjugated anti-rabbit IgG and a HRP substrate, the colors developed in proportion to the bound adiponectin, can be easily measured by Elisa plate reader.
Catalog No. 50-113-2327