Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AdipoR1 Goat anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
Goat Polyclonal Antibody has been used in 6 publications
$176.85 - $436.00
Specifications
Antigen | AdipoR1 |
---|---|
Concentration | 1.0 mg/mL |
Dilution | Western Blot 1:1000, Immunohistochemistry 1:250, Immunohistochemistry-Paraffin 1:250 |
Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Description
AdipoR1 Polyclonal specifically detects AdipoR1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
AdipoR1 | |
Western Blot 1:1000, Immunohistochemistry 1:250, Immunohistochemistry-Paraffin 1:250 | |
Polyclonal | |
Goat | |
Human, Mouse, Rat | |
ACDCR1, adiponectin receptor 1, adiponectin receptor protein 1, ADR1, CGI45, FLJ42464, PAQR1FLJ25385, Progestin and adipoQ receptor family member I, TESBP1A | |
ADIPOR1 | |
IgG | |
Affinity Purified |
1.0 mg/mL | |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
PBS (pH 7.2) and 1.0 mg/ml BSA with 0.1% Sodium Azide | |
51094 | |
Synthetic peptide (CYAARIPERFFPGKFDIWFQSHQIF) corresponding to aa 317-340 of the C-terminal domain of Human, Mouse and Rat ADPR 1 Receptor | |
Primary | |
Store at -80C. Avoid freeze-thaw cycles. |
For Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title