Learn More
AdipoR1 Rabbit anti-Human, Alexa Fluor™ 700, Clone: 2416C, R&D Systems™
Rabbit Monoclonal Antibody
Manufacturer:  R&D Systems FAB10128N100UG
Catalog No. FB10128N100
Add to cart
This item is not returnable. View return policy
Description
Description
AdipoR1 Monoclonal antibody specifically detects AdipoR1 in Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
AdipoR1
2416C
Flow Cytometry 0.25-1 ug/10^6 cells
Purified, Supernatant, Conjugated
Rabbit
IgG
100 μg
Primary
51094.0
Human
Flow Cytometry
Alexa Fluor™ 700
Supplied 0.2 mg/mL in a saline solution containing BSA and sodium azide.
ACDCR1, adiponectin receptor 1, adiponectin receptor protein 1, ADR1, CGI45, FLJ42464, PAQR1, PAQR1FLJ25385, Progestin and adipoQ receptor family member I, TESBP1A
Synthetic peptide containing human AdipoR1 C-term sequences,
Protein A or G purified from cell culture supernatant
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied.
Monoclonal
Detects human AdipoR1 in direct ELISAs. In direct ELISA, no cross-reactivity with human AdipoR2 is observed