Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AdipoR2 Goat anti-Human, Mouse, Polyclonal, Novus Biologicals™
Goat Polyclonal Antibody
$177.03 - $436.00
Specifications
Antigen | AdipoR2 |
---|---|
Concentration | 1.0 mg/mL |
Dilution | Western Blot 1:1000, Immunohistochemistry, Immunohistochemistry-Paraffin 1:250 |
Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Description
AdipoR2 Polyclonal specifically detects AdipoR2 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
AdipoR2 | |
Western Blot 1:1000, Immunohistochemistry, Immunohistochemistry-Paraffin 1:250 | |
Polyclonal | |
Goat | |
Human, Mouse | |
Q86V24 | |
79602 | |
Synthetic peptide (30 amino acid) CSRTPEPDIRLRKGHQLDGTRRGDNDSHQG corresponding to residues 11-40 of the N-terminal domain of human AdipoR2. | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
1.0 mg/mL | |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
10mM KHPO4, 0.14M NaCl and 1.0 mg/ml BSA, pH 7.2 with 0.1% Sodium Azide | |
ACDCR2, adiponectin receptor 2, adiponectin receptor protein 2, FLJ21432, PAQR2MGC4640, Progestin and adipoQ receptor family member II | |
ADIPOR2 | |
IgG | |
Affinity Purified |
For Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title