Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Adipoyl chloride, 98%, ACROS Organics™

Click to view available options
Quantity:
100mL
500mL
Packaging:
Glass bottle
111-50-2
C6H8Cl2O2
183.03
MFCD00000759
PWAXUOGZOSVGBO-UHFFFAOYSA-N
adipoyl chloride,adipoyl dichloride,adipyl chloride,adipoylchloride,adipic dichloride,hexanedioyl chloride,adipic acid dichloride,butane-1,4-dicarbonyl chloride,adipic acid chloride,pubchem13242
61034
hexanedioyl dichloride
C(CCC(=O)Cl)CC(=O)Cl
111-50-2
C6H8Cl2O2
183.03
MFCD00000759
PWAXUOGZOSVGBO-UHFFFAOYSA-N
adipoyl chloride,adipoyl dichloride,adipyl chloride,adipoylchloride,adipic dichloride,hexanedioyl chloride,adipic acid dichloride,butane-1,4-dicarbonyl chloride,adipic acid chloride,pubchem13242
61034
hexanedioyl dichloride
C(CCC(=O)Cl)CC(=O)Cl
Specific Gravity 1.25
Infrared Spectrum Authentic
Refractive Index 1.4700 to 1.4720
Flash Point 72°C
Quantity 500mL
Boiling Point 105.0°C to 107.0°C (2.0 mmHg)
Density 1.2500g/mL
Color Brown
Packaging Glass bottle
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula ClCO(CH2)4COCl
Beilstein 02, 653
Solubility Information Solubility in water: reacts. Other solubilities: forming hcl
Formula Weight 183.03
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material Adipoyl chloride, 98%
Show More Show Less

GHS H Statement
Causes serious eye damage.
Causes severe skin burns and eye damage.
Harmful in contact with skin.
Harmful if inhaled.
Harmful if swallowed.
Contact with water liberates toxic gas.
Reacts violentl

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately

GHS Signal Word: Danger

EINECSNumber : 203-876-4

TSCA : TSCA