Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Adjustable Height Heavy Duty Steel Table with Locking Swivel Casters, Phenolic Work Surface

Fisherbrand™ Heavy Duty Steel Tables are great for general lab use.

 View more versions of this product

Catalog No. 06-000-824


Add to cart

Description

Description

  • Comes with 4 locking swivel casters.
  • Steel frame is 2 x 2 in. tubular steel frame construction
  • Adjustable Height Heavy Duty Tables with casters are rated for 960 lbs UDL
Specifications

Specifications

960 lbs.
Fisherbrand White
91.4 cm
Fully Welded 2 in. x 2 in. Tubular Steel
Table Frame, Phenolic Work Surface and Locking Swivel Casters
48 in.
435.4 kg
36 in.
30 to 36 in.
76.2 to 91.4 cm
2 years
121.9 cm
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit