Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.

Diversified Woodcrafts™ Adjustable Wooden Leg Oak Table Products for Science Education Available on GSA/VA Contract for Federal Government customers only.

Legs adjust from sitting height of 30 in. to standing height of 36 in.

$650.00 - $873.00

Specifications

Includes Adjustable wooden legs
Size D x W: 24 x 60 in.
View More Specs

Products 4
Catalog Number Mfr. No. Top Style Price Quantity & Availability  
Catalog Number Mfr. No. Top Style Price Quantity & Availability  
S38068 Available on GSA/VA Contract for Federal Government customers only. Products for Science Education View Documents Diversified Woodcrafts™
A7601K
Plastic laminate Each for $650.00
N/A
S38070 Available on GSA/VA Contract for Federal Government customers only. Products for Science Education View Documents Diversified Woodcrafts™
A7604K
Phenolic Each for $809.00
N/A
S38069 Available on GSA/VA Contract for Federal Government customers only. Products for Science Education View Documents Diversified Woodcrafts™
A7602K
ChemGuard Each for $690.00
N/A
S38071 Available on GSA/VA Contract for Federal Government customers only. Products for Science Education View Documents Diversified Woodcrafts™
A7606K
Epoxy Each for $873.00
N/A
Description

Description

  • Solid oak aprons
  • Chemical-resistant finish
    • This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Specifications

Adjustable wooden legs
D x W: 24 x 60 in.
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit