Learn More
Alfa Aesar™ Adjuvant Peptide
For cellular and molecular biology applications
Manufacturer:  Alfa Aesar™ J62385LB0
 View more versions of this product
Catalog No. AAJ62385LB0
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Specifications
492.48(anhy)
5mg
Powder
RUO
Muramyl dipeptide
HPLC
Used in immunization to replace more complex proteins. Inhibits the HIV replication in CD4+ H9 lymphocytes
N-Acetylmuramyl-L-alanyl-D-isoglutamine
2° to 8°C
Adjuvant Peptide