Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ADK Antibody, Alexa Fluor™ 405, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP300077AF405
Description
ADK Polyclonal specifically detects ADK in Human samples. It is validated for Western Blot, ELISA.Specifications
ADK | |
Polyclonal | |
Western Blot, ELISA | |
adenosine kinase, AKAdenosine 5'-phosphotransferase, EC 2.7.1, EC 2.7.1.20 | |
Produced in rabbits immunized with purified, recombinant ADK (AAH03568.1; Met 1-His 345). | |
0.1 mL | |
Cancer, Cell Cycle and Replication, Immunology, Lipid and Metabolism | |
132 | |
Store at 4°C in the dark. | |
IgG |
Western Blot, ELISA | |
Alexa Fluor 405 | |
50mM Sodium Borate | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction