Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ADK Rabbit anti-Human, Alexa Fluor 700, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP300077AF700

Catalog No. NB064248


Add to cart

Description

Description

ADK Polyclonal antibody specifically detects ADK in Human samples. It is validated for Western Blot, ELISA
Specifications

Specifications

ADK
Alexa Fluor 700
50mM Sodium Borate
adenosine kinase, AKAdenosine 5'-phosphotransferase, EC 2.7.1, EC 2.7.1.20
Produced in rabbits immunized with purified, recombinant ADK (AAH03568.1; Met 1-His 345).
Affinity purified
RUO
Store at 4°C in the dark.
Polyclonal
Human
ELISA, Western Blot
Western Blot, ELISA
Purified
Rabbit
IgG
0.1 mL
Cancer, Cell Cycle and Replication, Immunology, Lipid and Metabolism
Primary
132
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit