Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Brady™ Admittance Sign: DANGER - HIGH VOLTAGE KEEP OUT
Identify electrical hazard and high voltage areas
Supplier: Brady™ 84082
View more versions of this product
Safety Services Available
Overview
Logo/Customization
Description
- Available in two materials - plastic and self sticking polyester
- Black/red on white
Specifications
ANSI Z53.1-1967 | |
Pressure-sensitive Acrylic | |
5 in. | |
DANGER HIGH VOLTAGE KEEP OUT | |
Red on White |
Polyester | |
Abrasion, Chemical | |
3.5 in. | |
Admittance Sign |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction